Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Reserpine certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018378 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8378 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8378 Supplier MilliporeSigma Supelco Supplier No. 0685950MG
Add to Cart
Add to Cart

Specifications

Product Line TraceCERT
CAS 50-55-5
Linear Formula C33H40N2O9
MDL Number MFCD00005091
Melting Point 265°C
Synonym (3 beta, 16 beta, 17 alpha, 18 beta, 20 alpha)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester
Type Certified Reference Material
Quantity 50 mg
Formula Weight 608.68
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.