Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TraceCERT™ Silicon Standard for ICP, 1,000 mg/L Si in Nitric Acid, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM. Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
Specifications
Specifications
| CAS | 7697-37-2 |
| Product Line | TraceCERT |
| Purity Grade Notes | 1000 mg/L Si in 2% HNO3 (contains HF traces), prepared with high purity Si, HNO3, HF and water |
| Concentration | 1000 mg/L Si in nitric acid |
| Type | Certified Reference Material |
| Quantity | 100 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?