Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Tamoxifen, MilliporeSigma™ Supelco™

Catalog No. 111012377 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2377 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2377 Supplier MilliporeSigma Supelco Supplier No. 0673450MG
Add to Cart
Add to Cart

Specifications

Product Line TraceCERT
CAS 10540-29-1
Linear Formula C6H5 C(C2H5)=C(C6H5)C6H4 OCH2 CH2 N(CH3)2
MDL Number MFCD00010454
Melting Point 97°C to 98°C
Synonym (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene; trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 371.51
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.