Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Tetrachloroethene Solution, 200 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020658 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0658 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0658 Supplier MilliporeSigma Supelco Supplier No. 48609

missing translation for 'validMainframeMsg'

Tetrachloroethene is a colorless liquid having an ethereal odor, which can find applications as a chemical intermediate and dry cleaning agent. It can also undergo reductive dehalogenation in the presence of an anaerobic bacterium to yield cis-1,2-dichloroethene.

Environmental and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 127-18-4
Flash Point 11°C
Synonym Tetrachloroethene solution; Perchloroethylene
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 200 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.