Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Thiacloprid certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018426 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8426 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8426 Supplier MilliporeSigma Supelco Supplier No. 1478350MG
Add to Cart
Add to Cart

Find more information here: Neonicotinoids

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 111988-49-9
Linear Formula C10H9 ClN4 S
MDL Number MFCD02101042
Synonym [3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidinylidene]cyanamide
Type Certified Reference Material
Quantity 50 mg
Formula Weight 252.72
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.