Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Thiamethoxam, MilliporeSigma™ Supelco™

Catalog No. 111018478 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8478 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8478 Supplier MilliporeSigma Supelco Supplier No. 6890150MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiamethoxam is a neonicotinoid broad spectrumand systemic insecticide, which belongs to the subclass of thianicotinyls. It can find applications in controlling the growth of sucking insects like aphids, whiteflies, and rice hoppers by acting on the acetylcholine nicotinic receptors.Find more information here: Neonicotinoids
TRUSTED_SUSTAINABILITY

Specifications

CAS 153719-23-4
Product Line TraceCERT
Linear Formula C8H10 ClN5O3 S
MDL Number MFCD03792862
Synonym 3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-N-nitro-4 H-1,3,5-oxadiazin-4-imine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 291.71
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.