Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Tin Standard for ICP, 1,000 mg/L Sn in Hydrochloric Acid, MilliporeSigma™ Supelco™

Catalog No. 111016930 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6930 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6930 Supplier MilliporeSigma Supelco Supplier No. 92615100ML
Add to Cart
Add to Cart
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM. Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-01-0
Product Line TraceCERT
Purity Grade Notes 1000 mg/L Sn in 10% hydrochloric acid, prepared with high purity tin metal, HCl and water
Concentration 1000 mg/L Sn in hydrochloric acid
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.