Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ TO11/IP-6A Aldehyde/Ketone-DNPH Mix, 15 μg/mL, MilliporeSigma™ Supelco™

Catalog No. 111016447 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6447 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6447 Supplier MilliporeSigma Supelco Supplier No. CRM4M7285
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

15 μg/mL Each Component in Acetonitrile (as the aldehyde or ketone)

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 15 μg/mL each component in acetonitrile (as the aldehyde or ketone)
Type Organic Pollutant Standard
Quantity 3 x 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Acetaldehyde-2,4-dinitrophenylhydrazone; Acetone-2,4-dinitrophenylhydrazone; Acrolein-2,4-dinitrophenylhydrazone; Benzaldehyde-2,4-dinitrophenylhydrazone; Butyraldehyde-2,4-dinitrophenylhydrazone; Crotonaldehyde-2,4-dinitrophenylhydrazone; 2,5-Dimethylbenzaldehyde 2,4-dinitrophenylhydrazone; Formaldehyde-2,4-dinitrophenylhydrazone; Hexaldehyde-2,4-dinitrophenylhydrazone; Isovaleraldehyde 2,4-dinitrophenylhydrazone; Propionaldehyde-2,4-dinitrophenylhydrazone; o-Tolualdehyde 2,4-dinitrophenylhydrazone; m-Tolualdehyde 2,4-dinitrophenylhydrazone; p-Tolualdehyde 2,4-dinitrophenylhydrazone; Valeraldehyde-2,4-dinitrophenylhydrazone

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.