Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ trans-9-Octadecenoic acid methyl ester, 10 mg/mL in heptane, 99.9%, MilliporeSigma™ Supelco™

Catalog No. 111020338 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0338 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0338 Supplier MilliporeSigma Supelco Supplier No. CRM46903
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 1937-62-8
Density 0.871 g/mL (at 20°C)
Flash Point -3.99°C - Closed Cup
Physical Form Liquid
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COOCH3
MDL Number MFCD00066521
Synonym Methyl elaidate; Elaidic acid methyl ester; Methyl trans-9-octadecenoate
Type Certified Reference Material
Quantity 1 mL
Formula Weight 296.49
Percent Purity 99.9%
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 10 mg/mL in heptane
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.