Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Tutin, MilliporeSigma™ Supelco™

Catalog No. 111018365 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8365 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8365 Supplier MilliporeSigma Supelco Supplier No. 807180.5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.Certified content incl. uncertainty and expiry date are given on the certificate.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2571-22-4
Product Line TraceCERT
Concentration 20 μg/g in acetonitrile (nominal concentration)
Linear Formula C15H18O6
Synonym (1 AS,1 bR,2 S,2′R,5 R,6 S,6 aR,7 aR,8 R)-Hexahydro-1 b,6-dihydroxy-6 A-methyl-8-(1-methylethenyl)spiro[2,5-methano-7 H-oxireno[3,4 ]cyclopent[1,2-d]oxepin-7,2′-oxiran]-3(2 H)-one solution
Type Certified Reference Material
Quantity 0.5 mL
Formula Weight 294.30
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.