Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Vanillin, MilliporeSigma™ Supelco™

Catalog No. 111012765 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2765 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2765 Supplier MilliporeSigma Supelco Supplier No. 3030450MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vanillin is an aromatic compound, and is used for flavoring foods. It is has biogenetic relationship to phenylpropanoid pathway. It is mostly used in foods, beverages, perfumes and pharmaceuticals.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Product Line TraceCERT
CAS 121-33-5
Vapor Pressure >0.01 mmHg (25°C)
Vapor Density 5.3 (vs. air)
Flash Point 153°C
Physical Form Neat
Linear Formula 4-(HO)C6H3-3-(OCH3)CHO
MDL Number MFCD00006942
Melting Point 81°C to 83°C
Boiling Point 170°C (15 mmHg)
Synonym 4-Hydroxy-3-methoxybenzaldehyde; Vanillic aldehyde
Type Certified Reference Material
Quantity 50 mg
Formula Weight 152.15
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.