Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Zearalenone Solution, 50 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111019240 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9240 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9240 Supplier MilliporeSigma Supelco Supplier No. CRM46916
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 17924-92-4
Flash Point 6°C - Closed Cup
Linear Formula C18H22O5
MDL Number MFCD00133085
Synonym ZON
Type Food and Cosmetic Component Standard
Quantity 1 mL
Formula Weight 318.36
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in acetonitrile
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.