Learn More
Andwin Scientific TRACECLEAN 20ML QA VIALS 72/CS
SDP
Catalog No. NC0563981
Encompass_Preferred
The TraceClean 20 mL Vials are constructed with Type I clear borosilicate glass. This product is ideal for use in various analyses and in specimen, extract, and standards storage. The vials are produced in a solvent-free facility to eliminate the ch
Catalog No. NC0563981 Supplier Andwin Scientific Supplier No. 89093860
The TraceClean 20 mL Vials are constructed with Type I clear borosilicate glass. This product is ideal for use in various analyses and in specimen, extract, and standards storage. The vials are produced in a solvent-free facility to eliminate the ch