Learn More
Glass Expansion Tracey Spray Chamber with Helix, 50ml cyclonic, Borosilicate glass. Includes 70-803-1439 Helix CT locking screw with seal (1 unit)
SDP
Catalog No. 501906917
Encompass_Preferred
Tracey spray chamber, includes Helix CT locking screw & seal (1 pc) & 13mm ball joint clip (1pc). Made from borosilicate glass, 50mL volume & has gravity drain. Compatible with Shimadzu ICPE 9000/9800 ICP-OES.The Tracey is a cyclonic design providing the highest sensitivity and fastest washout performance for ICP analyses. Ideal choice for most ICP applications, especially for those requiring the highest sample throughput and performance.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include NC2081825 (suggested to replace every 6 months) & JC-13.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6917 Supplier Glass Expansion Supplier No. 208088881HE
Tracey spray chamber, includes Helix CT locking screw & seal (1 pc) & 13mm ball joint clip (1pc). Made from borosilicate glass, 50mL volume & has gravity drain. Compatible with Shimadzu ICPE 9000/9800 ICP-OES.The Tracey is a cyclonic design providing the highest sensitivity and fastest washout performance for ICP analyses. Ideal choice for most ICP applications, especially for those requiring the highest sample throughput and performance.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include NC2081825 (suggested to replace every 6 months) & JC-13.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001