Learn More
Glass Expansion Tracey Spray Chamber with Helix CT
SDP
Catalog No. 502703183
Encompass_Preferred
Tracey Spray Chamber with Helix CT (N8151077). Includes UniFit with 1.6mm (1/16") OD x 0.75mm ID x 700mm long tube (1 pc) and Helix CT with Locking Screw and Seal (1 pc). Made from borosilicate glass, 50mL volume. Compatible with PerkinElmer NexION 5000 ICP-MS.The Tracey is a cyclonic design providing the highest sensitivity and fastest washout performance for ICP analyses. Ideal choice for most ICP applications, especially for those requiring the highest sample throughput and performance.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include NC1943061 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3183 Supplier Glass Expansion Supplier No. 208094419HE
Tracey Spray Chamber with Helix CT (N8151077). Includes UniFit with 1.6mm (1/16") OD x 0.75mm ID x 700mm long tube (1 pc) and Helix CT with Locking Screw and Seal (1 pc). Made from borosilicate glass, 50mL volume. Compatible with PerkinElmer NexION 5000 ICP-MS.The Tracey is a cyclonic design providing the highest sensitivity and fastest washout performance for ICP analyses. Ideal choice for most ICP applications, especially for those requiring the highest sample throughput and performance.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include NC1943061 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001