Learn More
Glass Expansion Tracey Spray Chamber with Helix CT 50ml cyclonic, Borosilicate glass. Includes UFT-075 UniFit Connector (1 unit) and 70-803-1439 Helix CT locking screw with seal (1 unit)
SDP
Catalog No. 501906946
Encompass_Preferred
Tracey spray chamber, includes Helix CT locking screw & seal (1 pc), UniFit with 1.3mm OD x 0.75mm ID x 700mm long tube (1 pc) & 13mm ball joint clip. Made from borosilicate glass, 50mL volume, fixed 90 degree outlet with 13mm ball joint. The Tracey is a cyclonic design providing the highest sensitivity and fastest washout performance for ICP analyses. Ideal choice for most ICP applications, especially for those requiring the highest sample throughput and performance.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include UFT-075 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6946 Supplier Glass Expansion Supplier No. 208092547HE
Tracey spray chamber, includes Helix CT locking screw & seal (1 pc), UniFit with 1.3mm OD x 0.75mm ID x 700mm long tube (1 pc) & 13mm ball joint clip. Made from borosilicate glass, 50mL volume, fixed 90 degree outlet with 13mm ball joint. The Tracey is a cyclonic design providing the highest sensitivity and fastest washout performance for ICP analyses. Ideal choice for most ICP applications, especially for those requiring the highest sample throughput and performance.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include UFT-075 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001