Learn More
Glass Expansion Tracey TFE Spray Chamber Base Cap
SDP
Catalog No. 501906967
Encompass_Preferred
Tracey TFE Spray Chamber Base Cap. A replacement TFE spray chamber cap for Glass Expansion’s Tracey TFE spray chambers.Replacement O-Rings can be ordered as V-044. The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6967 Supplier Glass Expansion Supplier No. 218092863
Tracey TFE Spray Chamber Base Cap. A replacement TFE spray chamber cap for Glass Expansion’s Tracey TFE spray chambers.Replacement O-Rings can be ordered as V-044. The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001