Learn More
Glass Expansion Tracey TFE Spray Chamber with B14 Drain and Helix CT, 50ml cyclonic, PTFE. Includes UFT-16-75 and UFT-2-700 UniFit Connectors (1 each) and 70-803-1439 Helix CT locking screw with seal (1 unit)
SDP
Catalog No. 501906942
Encompass_Preferred
Tracey PTFE spray chamber, includes Helix CT locking screw & seal and JC-13 Metal ball Joint Clip (1 ea). Made from PTFE, 50mL volume. Compatible with Shimadzu ICPE-9000/9800 & ICPS-7500/8100 ICP-OES.The Tracey PTFE is a HF-resistant cyclonic design with a proprietary Stediflow surface treatment that significantly improves sensitivity & precision, in addition to enhancing washout efficiency compared to other inert spray chamber designs. The interior surface can be refurbished at a nominal cost, average lifespan of treatment is 12 months when well cared for.The main feature of the Helix CT interface is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume & chemically inert seal of the nebulizer.Replacement parts include NC2081825 & V-044, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6942 Supplier Glass Expansion Supplier No. 208092508
Tracey PTFE spray chamber, includes Helix CT locking screw & seal and JC-13 Metal ball Joint Clip (1 ea). Made from PTFE, 50mL volume. Compatible with Shimadzu ICPE-9000/9800 & ICPS-7500/8100 ICP-OES.The Tracey PTFE is a HF-resistant cyclonic design with a proprietary Stediflow surface treatment that significantly improves sensitivity & precision, in addition to enhancing washout efficiency compared to other inert spray chamber designs. The interior surface can be refurbished at a nominal cost, average lifespan of treatment is 12 months when well cared for.The main feature of the Helix CT interface is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume & chemically inert seal of the nebulizer.Replacement parts include NC2081825 & V-044, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001