Learn More
TARGETMOL CHEMICALS INC TRACHELOSIDE 10MG
SDP
Catalog No. 502580104
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tracheloside significantly decreases the activity of alkaline phosphatase (IC50 0.31 ug/ml) a level of inhibition comparable to that of tamoxifen (IC50 0.43 ug/ml). purity: 99%
Catalog No. 50-258-0104 Supplier TARGETMOL CHEMICALS INC Supplier No. T385510MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tracheloside significantly decreases the activity of alkaline phosphatase (IC50 0.31 ug/ml) a level of inhibition comparable to that of tamoxifen (IC50 0.43 ug/ml). purity: 99%