Learn More
Medchemexpress LLC Tracheloside | 33464-71-0 | 550.55 | 5 MG
SDP
Catalog No. 5000415653
Encompass_Preferred
Tracheloside is an antiestrogenic lignin that promotes keratinocyte proliferation through ERK1/2 stimulation. It is considered a good candidate to promote wound healing.
  • Antiestrogenic lignin
  • Promotes keratinocyte proliferation through ERK1/2 stimulation
  • Good candidate to promote wound healing
Catalog No. 50-004-15653 Supplier Medchemexpress LLC Supplier No. HYN15075MG
Tracheloside is an antiestrogenic lignin that promotes keratinocyte proliferation through ERK1/2 stimulation. It is considered a good candidate to promote wound healing.
  • Antiestrogenic lignin
  • Promotes keratinocyte proliferation through ERK1/2 stimulation
  • Good candidate to promote wound healing