Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tracheloside | 33464-71-0 | MFCD00870631 | 99.8% | 550.55 g·mol⁻¹ | C27H34O12 | 10 MG
SDP

Catalog No. 5000296677
Encompass_Preferred

Tracheloside is a plant-derived lignan supplied as a white to off-white solid for research applications. It shows antiestrogenic activity and promotes keratinocyte proliferation via ERK1/2 stimulation, making it a candidate for wound-healing research.

  • High purity (99.8%).
  • Molecular weight 550.55 g·mol⁻¹.
  • Soluble in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Stable as a solid at 4°C; protect from light.
  • Provided with datasheet and certificate of analysis for quality verification.

Catalog No. 50-002-96677 Supplier Medchemexpress LLC Supplier No. HYN150710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tracheloside is a plant-derived lignan supplied as a white to off-white solid for research applications. It shows antiestrogenic activity and promotes keratinocyte proliferation via ERK1/2 stimulation, making it a candidate for wound-healing research.

  • High purity (99.8%).
  • Molecular weight 550.55 g·mol⁻¹.
  • Soluble in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Stable as a solid at 4°C; protect from light.
  • Provided with datasheet and certificate of analysis for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.