Learn More
MSC Honeywell MTS Track Safety Shoe, Mid-Cut, Composite Toe, 1 ea
Catalog No. 5000507580
Encompass
The MTS Track S3S is a mid-cut, sporty safety shoe engineered for performance and ergonomics in logistics, services, and transportation environments. Featuring a high-tech design inspired by professional athletic footwear, this shoe is constructed with durable, breathable materials to provide protection without compromising comfort.
  • Composite toe cap provides protection against impacts up to 200 Joules
  • Anti-puncture non-metallic midsole (type PS) for underfoot safety
  • Slip-resistant outsole conforms to the SR standard
  • Certified to EN ISO 20345:2022 S3S SR FO HI CI ESD
  • Integrated TPU heel counter for enhanced support and flexibility
  • Thermoformed, anatomical, antistatic polyurethane foam insock
  • Compatible with DGUV 112-191 certified orthopedic insoles
  • Water-resistant, breathable upper material
  • Heat and cold insulated sole with fuel oil resistance
Catalog No. 50-005-07580 Supplier MSC Supplier No. 79522694
The MTS Track S3S is a mid-cut, sporty safety shoe engineered for performance and ergonomics in logistics, services, and transportation environments. Featuring a high-tech design inspired by professional athletic footwear, this shoe is constructed with durable, breathable materials to provide protection without compromising comfort.
  • Composite toe cap provides protection against impacts up to 200 Joules
  • Anti-puncture non-metallic midsole (type PS) for underfoot safety
  • Slip-resistant outsole conforms to the SR standard
  • Certified to EN ISO 20345:2022 S3S SR FO HI CI ESD
  • Integrated TPU heel counter for enhanced support and flexibility
  • Thermoformed, anatomical, antistatic polyurethane foam insock
  • Compatible with DGUV 112-191 certified orthopedic insoles
  • Water-resistant, breathable upper material
  • Heat and cold insulated sole with fuel oil resistance