Learn More
MSC MTS Track Mid-Cut Sporty Workshoe, Ondura Nilo, S3S, 1 ea
Catalog No. 5000507564
Encompass
This mid-cut safety shoe features a casual, sporty design suitable for logistics, warehousing, transportation, and service industries. It provides reliable protection with a non-metallic composite toe cap and anti-perforation midsole, ensuring comfort through a lightweight construction and an adaptable fit system.
  • Composite toe cap protects against impact and compression
  • Dual-density injected PU outsole is oil resistant, slip resistant, and silicone-free
  • Modular three width insock fitting system allows for standard, wide, or narrow customization
  • TPU heel counter provides increased support, flexibility, and comfort
  • Water-resistant and breathable Ondura Nilo upper
  • Meets ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards
  • Features ESD and antistatic properties with heat and cold insulation
Catalog No. 50-005-07564 Supplier MSC Supplier No. 79509170
This mid-cut safety shoe features a casual, sporty design suitable for logistics, warehousing, transportation, and service industries. It provides reliable protection with a non-metallic composite toe cap and anti-perforation midsole, ensuring comfort through a lightweight construction and an adaptable fit system.
  • Composite toe cap protects against impact and compression
  • Dual-density injected PU outsole is oil resistant, slip resistant, and silicone-free
  • Modular three width insock fitting system allows for standard, wide, or narrow customization
  • TPU heel counter provides increased support, flexibility, and comfort
  • Water-resistant and breathable Ondura Nilo upper
  • Meets ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards
  • Features ESD and antistatic properties with heat and cold insulation