Learn More
EDVOTEK™ Tracking Health Through Urinalysis: A Case-Based Clinical Simulation

Shop All EDVOTEK Products


Description
- Students will analyze simulated urine samples using real world techniques utilized in biomedical labs.
- Learn to count identify white blood cells and crystals within samples.
- Perform diagnostic testing to reveal information about patient urine samples.
- Determine a diagnosis and treatment plan based on what ails each patient.
- For Up to 10 Lab Groups.
- Includes : Instructions, Patient Synthetic Urine Samples, pH Additive, Calcium Additive, 15 mL Conical Tubes, Urine Dipsticks, Microscope Slides, Small Transfer Pipets, Inoculating Loops
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Time Range | 45 Min. |
| Class Size | 10 Lab Groups |
| Product Type | Artificial Urine Control |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.