Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrackIt™ 1 Kb Plus DNA Ladder
GREENER_CHOICE

Catalog No. 10488085
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-488-085 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-488-085 Supplier Invitrogen™ Supplier No. 10488085
Only null left
Add to Cart
Edge
Add to Cart

Includes

TrackIt™ 1 Kb Plus DNA Ladder (0.1μg/μL) in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.6% tartrazine, and 5% glycerol; For 100 applications

Suitable for sizing linear double-stranded DNA fragments from 100 bp to 12 kb

TrackIt™ 1Kb Plus DNA Ladder is prepared from plasmid containing repeats of 1,000 bp DNA fragment. The 1,650bp band is generated from pUC and all bands <1,000 bp are from lambda DNA. TrackIt 1Kb Plus DNA Ladder is formulated with two unique tracking dyes, Xylene Cyanol FF (XCFF) and tartrazine, that allow to visually track DNA migration during electrophoresis and also indicate when maximum resolution is achieved. Tracking dyes do not obscure visualization of DNA bands in ladder as dyes run outside limits of most DNA bands in ladder.

  • Consists of 12 bands in 1,000bp increments ranging in size from 1 to 12 Kb, a 1,650 bp fragment, and vector DNA fragments from 100 to 850 bp
  • Orientation doublet at 1,650 bp and 2,000 bp for easy reference
  • Formulated with unique tracking dyes, XCFF and tartrazine
  • Designed for use with E-Gel™ agarose gels and TBE/TAE agarose gels
  • Supplied in ready-to-load format, stable at room temperature
  • Visualized with ethidium bromide or SYBR™ Green staining
  • Compatibile with agarose gels E-Gel™ gels

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 500 μL TrackIt 1 Kb Plus DNA Ladder
• 1 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 24 months. For longer periods, store at -20°C.
Concentration 0.1 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 100 to 15000 bp
Volume (Metric) 500 μL
Gel Compatibility Agarose gel
No. of Reactions 100 Applications
Quantity 50 μg
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Line TrackIt
Product Type DNA Ladder
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.