Learn More
Gilson™ TRACKMANTM Connected
Catalog No. FB1022G Shop All Gilson Products
Change view
Click to view available options
TRACKMANTM Connected; Plug Type: US
Catalog No.
FB1022G TRACKMANTM Connected; Plug Type: US
1 options
Catalog No. FB1022G Supplier Gilson™ Supplier No. FB1022
Add to Cart
Add to Cart
TRACKMANTM Connected is a tablet with exclusive accessories and applications that make pipetting on 96- and 384-well plates faster and more reliable, improving efficiency at the bench by tracking pipetting tasks.
  • Add traceability in your experiment
  • Easily capture notes, photos, and monitor your protocols
  • Streamline your work with the Bluetooth-connected environment sensor by automatically registering a protocol run action such as pipetting and conditions like temperature, humidity, and air pressure
  • Boost your experiment reliability
  • When paired with PIPETMANTM M Connected, the installed PipettePilot app guides you through your microplate method
  • The app displays where, when, and how much to pipette with real-time interactions to prevent errors
  • Get immediate reproducibility
  • Effortlessly create and share your protocols
  • Compatibility with sciNote lets you structure your work into projects, experiments, and tasks, making it easy to visualize your work progress
  • Sync your results for a comprehensive view of your pipetting actions, samples, and protocols
  • Access pipette analytics and calibration history
  • PipetteScopeTM informs you exactly when your pipette needs to be calibrated
  • Through the app, you can easily request a calibration appointment and also view service history reports in one central location
  • PipetteScope gives you instant access to the pipette usage analytics as well as access to the user guide
Specifications
Battery Type Included
Material Polypropylene
Model TRACKMAN Connected
Display Pipette Pilot
For Use With (Equipment) PIPETMAN™ M Connected
Includes User's Guide
Plug Type US
Quantity 1
Width (English) 9.02 in