Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 54MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: ACP5/1070. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HepG2, 293T, K562 or RPMI-8226 Cells. Spleen from Hairy Cell Leukemia (HCL) patient. Cellular Staining: Cytoplasmic (Lysosomes). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human ACP5 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 35kDa.Chromosome: 19p13.2. Gene Symbol: ACP5. Gene ID: 54. Ensemble: ENSG00000102575. Uniprot: NP_001602.
Catalog No. 50-188-5072