Learn More
Aviva Systems Biology Corp TRAF1 Antibody - middle region (AVARP09008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRAF1. Alternate names: EBI6, MGC:10353ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-62562