Learn More
Enzo Life Sciences TRAF1 monoclonal antibody (3D4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM421E
Encompass_Preferred
Clone: 3D4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant mouse TRAF1. UniProt ID: P39428. Species reactivity: Mouse. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol. No preservatives.
Catalog No. 50-200-7144