Learn More
Aviva Systems Biology Corp TRAF3IP2 Antibody - N-terminal region (ARP52863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAF3IP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TRAF3IP2. Alternate names: ACT1, C6orf2, C6orf4, C6orf5, C6orf6, CIKS, DKFZp586G0522, MGC3581, PSORS13ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-777