Learn More
Aviva Systems Biology Corp TRAF3IP3 Antibody - C-terminal region (ARP49943_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49943_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAF3IP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRAF3IP3. Alternate names: DJ434O14.3, FLJ44151, MGC117354, MGC163289, T3JAMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53610