Learn More
Enzo Life Sciences TRAF6 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADIAAP426E
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant mouse TRAF6 (N-terminal fragment). UniProt ID: P70196. Species reactivity: Mouse. Applications: IP, WB. Recommended Dilutions: Immunoprecipitation (2-4µL/100-500 µl cell extract from 5x106 cells) Western Blot (1:1000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7164