Learn More
Aviva Systems Biology Corp TRAFD1 Antibody - C-terminal region (ARP32828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAFD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TRAFD1. Alternate names: FLN29ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52877