Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Traffic Barrel 41-1/2 in H 9 l

Catalog No. 502768980
Encompass_Preferred

Traffic Barrel 41-1/2 in H 9 lb.

Catalog No. 50-276-8980 Supplier Grainger Supplier No. 13P896
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Traffic Barrel 41-1/2 in H 9 lb.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.