Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Traffic Sign

Catalog No. 19169590 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
BE PREPARED TO STOP
EMERGENCY MEDICAL CARE
H (Hospital Symbol)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-169-590 H (Hospital Symbol)
19-169-587 BE PREPARED TO STOP
19-169-589 EMERGENCY MEDICAL CARE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-169-590 Supplier Accuform™ Supplier No. FRG110RA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accuform™ Traffic Signs safely direct and regulate the flow of vehicles and pedestrians around parking areas, dock areas, and roadways.

  • Durable, .080 in. aluminum signs will not rust
  • Engineer-Grade Prismatic Reflective sign face provides visibility in low-light conditions
  • UV-resistant inks prevent fading from long-term outdoor use
  • Clear UV Protective Overcoat
  • Post mount holes
  • Meets ASTM D 4956 Type I

Specifications

Certifications/Compliance ASTM D 4956 Type I
Grade Engineer
Material Reflective Aluminum
Mounting Type Corner Holes
Languages English
Dimensions (L x W) 24 x 24 in.
Displayed Text H (Hospital Symbol)
Color White on Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.