Learn More
Medchemexpress LLC Tragacanth gum | 9000-65-1 | MFCD00131255 | ≈840,000 g/mol (heterogeneous polymer; no single defined MW) | No single chemical formula (heterogeneous polysaccharide mixture) | 5 G
SDP
Catalog No. 5000281384
Encompass_Preferred
Tragacanth gum is a natural anionic polysaccharide supplied as a solid biochemical reagent. It is used as a thickening and stabilizing agent, and can act as a reducing agent in green synthesis of metallic nanoparticles. Reported bioactivities include nephroprotective effects and support for cell adhesion. For research use only.
  • Natural anionic polysaccharide derived from plant exudate.
  • High-molecular-weight polymer that acts as a stabilizer and thickener.
  • Can serve as a reducing agent and stabilizer in nanoparticle synthesis.
  • Reported bioactivities include nephroprotective, cell adhesion support, and anticoagulant potential.
  • Provided as a solid powder with SDS and COA available for documentation.
Catalog No. 50-002-81384 Supplier Medchemexpress LLC Supplier No. HYW2501215G
Tragacanth gum is a natural anionic polysaccharide supplied as a solid biochemical reagent. It is used as a thickening and stabilizing agent, and can act as a reducing agent in green synthesis of metallic nanoparticles. Reported bioactivities include nephroprotective effects and support for cell adhesion. For research use only.
  • Natural anionic polysaccharide derived from plant exudate.
  • High-molecular-weight polymer that acts as a stabilizer and thickener.
  • Can serve as a reducing agent and stabilizer in nanoparticle synthesis.
  • Reported bioactivities include nephroprotective, cell adhesion support, and anticoagulant potential.
  • Provided as a solid powder with SDS and COA available for documentation.