Learn More
Medchemexpress LLC Tragacanth gum | 9000-65-1 | MFCD00131255 | 840,000 g/mol (reported range 310,000-840,000) | Not applicable - complex polysaccharide mixture (no single chemical formula) | 10 G
SDP
Catalog No. 5000280523
Encompass_Preferred
Tragacanth gum is a natural anionic polysaccharide provided as a white to light-yellow powder for research applications. It is used as a biochemical assay reagent and excipient, with water solubility of ≥ 20 mg/mL, and can improve cell adhesion, exhibit anticoagulant activity in assays, and act as a stabilizer and reducing agent in green synthesis of metallic nanoparticles. Recommended storage varies by form to preserve stability.
Catalog No. 50-002-80523 Supplier Medchemexpress LLC Supplier No. HYW25012110G
Tragacanth gum is a natural anionic polysaccharide provided as a white to light-yellow powder for research applications. It is used as a biochemical assay reagent and excipient, with water solubility of ≥ 20 mg/mL, and can improve cell adhesion, exhibit anticoagulant activity in assays, and act as a stabilizer and reducing agent in green synthesis of metallic nanoparticles. Recommended storage varies by form to preserve stability.