Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tragacanth Gum MP Biomedicals

Catalog No. ICN10479280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10479280 100 g
ICN10479290 500 g
ICN10479291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10479280 Supplier MP Biomedicals, Inc Supplier No. 0210479280
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9000-65-1
MDL Number MFCD00131255
Synonym Gum tragacanth

Specifications

Color Cream
Quantity 100 g
Physical Form Powder
Chemical Name or Material Tragacanth Gum, Molecular biology and Cell Biology
EINECSNumber 232-552-5
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.