Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tragacanth, Powder, NF, Spectrum™ Chemical
SDP

Catalog No. 18607475 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-475 500 g Poly Bottle
18-607-478 50 kg Fiber Drum
18-607-476 2.5 kg Poly Pail
18-607-477 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-607-475 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR105500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

TR105, 9000-65-1

Spectrum™ Chemical Tragacanth, Powder, NF is used as an emulsifier, thickener, stabilizer, and texturant additive. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-65-1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.