Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tragacanth, Powder, NF, Spectrum™ Chemical
SDP

Catalog No. 18607474 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-474 125 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-474 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR105125GM

missing translation for 'validMainframeMsg'

TR105, 9000-65-1

Spectrum™ Chemical Tragacanth, Powder, NF is used as an emulsifier, thickener, stabilizer, and texturant additive. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-65-1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 125 g
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Tragacanth, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.