Learn More
Medchemexpress LLC HY-P7306 50ug Medchemexpress, TRAIL/Apo2L, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP730650UG
Encompass_Preferred
Medchemexpress, HY-P7306 50ug TRAIL/Apo2L, Human CAS: Recombinant Human TRAIL/Apo2L is a protein functioning as a ligand that induces the process of apoptosis through engagement of death receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4377