Learn More
Enzo Life Sciences TRAIL (human) monoclonal antibody (2E5) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804296C100
Encompass_Preferred
Clone: 2E5. Isotype: IgG1. Immunogen: Recombinant human soluble TRAIL (aa 95-281). Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Protein A-affinity purified. Sterile filtered. Formulation: Liquid. In PBS, pH 7.4. Contains no preservatives.
Catalog No. 50-200-9859