Learn More
Enzo Life Sciences TRAIL-(human) monoclonal antibody (RIK-2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049070100
Encompass_Preferred
Clone: RIK-2. Isotype: IgG1. Immunogen: Human TRAIL transfected mouse cell line. Species reactivity: Human. Applications: Flow Cytometry. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-0018