Learn More
Enzo Life Sciences TRAIL-(human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210732R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human soluble TRAIL (aa 95-281). UniProt ID: P50591. Source: Purified from serum. Species reactivity: Human. Specificity: Recognizes soluble TRAIL. Creossreactivity: Does not cross-react with mouse TRAIL. Applications: Flow Cytometry, ICC, IHC (FS), WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8145