Learn More
Enzo Life Sciences TRAIL-R1 (human) monoclonal antibody (DJR1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX8049120100
Encompass_Preferred
Clone: DJR1. Isotype: IgG1. Immunogen: Recombinant human TRAIL-R1 extracellular domain IgG1 Fc fusion protein. Species reactivity: Human. Applications: Flow Cytometry, WB. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-0020