Learn More
AdipoGen TRAIL-R1 to -R4 h FACS Pack
SDP
Catalog No. 502562752
Encompass_Preferred
Set. TRAIL-R1, -R2, -R3 and -R4 are receptors for the cytotoxic ligand TRAIL. When signaling through TRAIL-R1 and -R2, the adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex DISC performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases aspartate-specific cysteine proteases mediating apoptosis. They promote the activation of NF-kappaB. TRAIL-R3 lacks a cytoplasmic death domain, and TRAIL-R4 contains a truncated death domain, and hence are not capable of inducing apoptosis. They protect cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.
Catalog No. 50-256-2752 Supplier AdipoGen Supplier No. AG44B0004KI01
Set. TRAIL-R1, -R2, -R3 and -R4 are receptors for the cytotoxic ligand TRAIL. When signaling through TRAIL-R1 and -R2, the adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex DISC performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases aspartate-specific cysteine proteases mediating apoptosis. They promote the activation of NF-kappaB. TRAIL-R3 lacks a cytoplasmic death domain, and TRAIL-R4 contains a truncated death domain, and hence are not capable of inducing apoptosis. They protect cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.