Learn More
AdipoGen TRAIL-R2 (human):Fc (human) (rec.) (3 x 50 µg)
SDP
Supplier:  AdipoGen AG40B00713050
Encompass_Preferred
Protein. The extracellular domain of human TRAIL-R2 (aa 52-212) is fused to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse TRAIL. Purity: >95% (SDS-PAGE). TRAIL-R2 (DR5; CD262) is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.
Catalog No. 50-205-3128