Learn More
AdipoGen anti-TRAIL-R2 (human), mAb (HS201) (preservative free) (100 µg)
SDP
Supplier:  AdipoGen AG20B0023PFC100
Encompass_Preferred
Monoclonal Antibody. Recognizes human TRAIL-R2. Does not cross-react with human TRAIL-R1, -R3 or -R4. Isotype: Mouse IgG1. Clone: HS201. Applications: FACS, FUNC (Blocking), ICC, IP. Liquid. In PBS containing 10% glycerol. TRAIL-R2 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.
Catalog No. 50-205-2927