Learn More
AdipoGen anti-TRAIL-R2 (human), mAb (TR2.21) (100 µg)
SDP
Supplier:  AdipoGen AG20B0028C100
Encompass_Preferred
anti-TRAIL-R2 (human), mAb (TR2.21) (100 µg), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes human TRAIL-R2. Does not cross-react with human TRAIL-R1, -R3 or -R4. Isotype: Mouse IgG1. Clone: TR2.21. Applications: FACS, IHC. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. TRAIL-R2 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.
Catalog No. 50-148-9645