Learn More
Enzo Life Sciences TRAIL-R2 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522005C050
Encompass_Preferred
Alternative name: DR5, TNFRSF 10B, KILLER, CD262. Concentration: 1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human TRAIL-R2 (DR5) (aa 52-212) is fused to the Fc portion of human IgG1. Specificity: Binds human and mouse TRAIL. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9359